siRNA Details
| Gene Symbol: OR13C9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001956.1 | NP_001001956.1 | 1 | 53 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR13C9| Gene Symbol: OR13C9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001956.1 | NP_001001956.1 | 1 | 53 | ||