siRNA Details
| Gene Symbol: OR13D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004484.1 | NP_001004484.1 | 1 | 725 | ||
| GenBank mRNA | BC140916.1 | - | 1 | 768 | 9056728 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR13D1| Gene Symbol: OR13D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004484.1 | NP_001004484.1 | 1 | 725 | ||
| GenBank mRNA | BC140916.1 | - | 1 | 768 | 9056728 | |