siRNA Details
| Gene Symbol: YIPF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173834.3 | NP_776195.2 | 2, 3 | 475 | ||
| XM_011530923.2 | XP_011529225.1 | 2, 3 | 883 | |||
| XM_011530924.1 | XP_011529226.1 | 2, 3 | 257 | |||
| XM_017029450.1 | XP_016884939.1 | 2, 3 | 257 | |||
| GenBank mRNA | AK095555.1 | - | 2, 3 | 257 | ||
| AK292835.1 | - | 2, 3 | 217 | |||
| BC012469.1 | - | 2, 3 | 213 | 4452699 | ||

YIPF6
Genomic Map