siRNA Details
| Gene Symbol: YIPF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195214.1 | NP_001182143.1 | 6 | 802 | ||
| NM_173834.3 | NP_776195.2 | 7 | 931 | |||
| XM_011530924.1 | XP_011529226.1 | 7 | 713 | |||
| GenBank mRNA | AK024673.1 | - | 3 | 237 | ||
| AK292835.1 | - | 7 | 673 | |||
| AK303992.1 | - | 6 | 532 | |||
| BC012469.1 | - | 7 | 669 | 4452699 | ||

YIPF6
Genomic Map