siRNA Details
| Gene Symbol: NPEPPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006310.3 | NP_006301.3 | 21 | 2761 | ||
| XM_011525496.1 | XP_011523798.1 | 22 | 2563 | |||
| XM_017025372.1 | XP_016880861.1 | 21 | 2723 | |||
| XM_017025373.1 | XP_016880862.1 | 22 | 2563 | |||
| GenBank mRNA | AJ132583.1 | - | 21 | 2491 | ||
| AK096491.1 | - | 6 | 1219 | |||
| AK293450.1 | - | 15 | 1693 | |||
| AK293995.1 | - | 14 | 1675 | |||
| AK296887.1 | - | 20 | 2404 | |||
| AK297107.1 | - | 20 | 2265 | |||
| AK303037.1 | - | 22 | 2556 | |||
| BC065294.1 | - | 21 | 2450 | 6059589 | ||

NPEPPS
Genomic Map