siRNA Details
| Gene Symbol: FANCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004629.1 | NP_004620.1 | 12, 13 | 2134 | ||
| GenBank mRNA | AJ007669.1 | - | 12, 13 | 2134 | ||
| AK293427.1 | - | 12, 13 | 1672 | |||
| AK311348.1 | - | 11, 12 | 1608 | |||
| AK312987.1 | - | 12, 13 | 2059 | |||
| BC000032.2 | - | 12, 13 | 1780 | 3507258 | ||
| BC011623.2 | - | 12, 13 | 1907 | 4053144 | ||
| BT009813.1 | - | 12, 13 | 1642 | |||
| KU159422.1 | - | 12, 13 | 2147 | |||
| KU178053.1 | - | 12, 13 | 1642 | |||
| KU178054.1 | - | 12, 13 | 1624 | |||
| U70310.1 | - | 12, 13 | 1776 | |||

FANCG
Genomic Map