siRNA Details
| Gene Symbol: KRT73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175068.2 | NP_778238.1 | 8 | 1385 | ||
| XM_011538257.2 | XP_011536559.1 | 7 | 2863 | |||
| GenBank mRNA | AJ508776.1 | - | 8 | 1385 | ||
| BC109212.2 | - | 7 | 1743 | 40007200 | ||

KRT73
Genomic Map