siRNA Details
| Gene Symbol: GALC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000153.3 | NP_000144.2 | 5 | 603 | ||
| NM_001201401.1 | NP_001188330.1 | 4 | 534 | |||
| NM_001201402.1 | NP_001188331.1 | 5 | 584 | |||
| XM_011536618.2 | XP_011534920.1 | 5 | 885 | |||
| GenBank mRNA | AK296530.1 | - | 4 | 425 | ||
| AK302683.1 | - | 5 | 757 | |||
| AK302956.1 | - | 5 | 423 | |||
| BC036518.2 | - | 5 | 455 | 5267705 | ||
| D25283.1 | - | 5 | 486 | |||
| D25284.1 | - | 5 | 486 | |||
| DA247447.1 | - | 5 | 527 | |||
| DA332567.1 | - | 5 | 567 | |||
| DC334180.1 | - | 5 | 584 | |||
| L23116.1 | - | 5 | 495 | |||

GALC
Genomic Map