siRNA Details
| Gene Symbol: MYBPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000256.3 | NP_000247.2 | 33 | 3695 | ||
| GenBank mRNA | AK291569.1 | - | 33 | 3695 | ||
| BC136543.1 | - | 33 | 3672 | 9020533 | ||
| BC136546.1 | - | 33 | 3672 | 9020536 | ||
| BC142685.1 | - | 33 | 3675 | 40148564 | ||
| BC151211.1 | - | 33 | 3678 | 40147517 | ||
| EF560722.1 | - | 33 | 3696 | |||
| EU747718.1 | - | 33 | 3695 | |||
| EU747719.1 | - | 33 | 3695 | |||

MYBPC3
Genomic Map