siRNA Details
| Gene Symbol: ZAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175619.2 | NP_783318.1 | 4 | 1195 | ||
| GenBank mRNA | AY191416.1 | - | 4 | 1155 | ||
| BC136265.1 | - | 4 | 1195 | 9020252 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZAR1| Gene Symbol: ZAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175619.2 | NP_783318.1 | 4 | 1195 | ||
| GenBank mRNA | AY191416.1 | - | 4 | 1155 | ||
| BC136265.1 | - | 4 | 1195 | 9020252 | ||