siRNA Details
| Gene Symbol: MFN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127660.1 | NP_001121132.1 | 9 | 1333 | ||
| NM_014874.3 | NP_055689.1 | 10 | 1478 | |||
| XM_005263543.3 | XP_005263600.1 | 11 | 1491 | |||
| XM_005263545.3 | XP_005263602.1 | 10 | 1341 | |||
| XM_005263547.3 | XP_005263604.1 | 10 | 1373 | |||
| XM_005263548.3 | XP_005263605.1 | 11 | 1508 | |||
| GenBank mRNA | AF036536.2 | - | 10 | 1347 | ||
| AK021947.1 | - | 4 | 513 | |||
| AK289828.1 | - | 9 | 1278 | |||
| AK295890.1 | - | 8 | 805 | |||
| AY028429.1 | - | 9 | 1051 | |||
| BC017061.1 | - | 10 | 1208 | 3901235 | ||
| D86987.1 | - | 10 | 1347 | |||

MFN2
Genomic Map