siRNA Details
| Gene Symbol: MFN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127660.1 | NP_001121132.1 | 2 | 335 | ||
| NM_014874.3 | NP_055689.1 | 3 | 480 | |||
| XM_005263543.3 | XP_005263600.1 | 4 | 493 | |||
| XM_005263545.3 | XP_005263602.1 | 3 | 343 | |||
| XM_005263547.3 | XP_005263604.1 | 3 | 375 | |||
| XM_005263548.3 | XP_005263605.1 | 4 | 510 | |||
| GenBank mRNA | AF036536.2 | - | 3 | 349 | ||
| AK021947.1 | - | 3 | 349 | |||
| AK289828.1 | - | 2 | 280 | |||
| AK295890.1 | - | 3 | 215 | |||
| AY028429.1 | - | 2 | 53 | |||
| BC017061.1 | - | 3 | 210 | 3901235 | ||
| BU178912.1 | - | 3 | 275 | |||
| D86987.1 | - | 3 | 349 | |||
| DC401346.1 | - | 3 | 480 | |||

MFN2
Genomic Map