siRNA Details
| Gene Symbol: POMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013382.5 | NP_037514.2 | 12 | 1472 | ||
| XM_011536675.2 | XP_011534977.1 | 12 | 1472 | |||
| XM_011536676.2 | XP_011534978.1 | 11 | 1385 | |||
| XM_011536677.2 | XP_011534979.1 | 8 | 1014 | |||
| XM_011536679.2 | XP_011534981.1 | 8 | 674 | |||
| XM_017021244.1 | XP_016876733.1 | 11 | 1306 | |||
| XM_017021245.1 | XP_016876734.1 | 8 | 760 | |||
| GenBank mRNA | AB209114.1 | - | 12 | 1294 | ||
| AF105020.1 | - | 12 | 1475 | |||
| AK023804.1 | - | 6 | 2063 | |||
| AK025962.1 | - | 2 | 91 | |||
| AY090480.1 | - | 12 | 1475 | |||
| BC031651.1 | - | 12 | 1362 | 5169145 | ||
| BM127609.1 | - | 6 | 396 | |||
| CR627447.1 | - | 12 | 1042 | |||

POMT2
Genomic Map