siRNA Details
| Gene Symbol: POMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013382.5 | NP_037514.2 | 11 | 1436 | ||
| XM_011536675.2 | XP_011534977.1 | 11 | 1436 | |||
| XM_011536676.2 | XP_011534978.1 | 10 | 1349 | |||
| XM_011536677.2 | XP_011534979.1 | 7 | 978 | |||
| XM_011536679.2 | XP_011534981.1 | 7 | 638 | |||
| XM_017021244.1 | XP_016876733.1 | 10 | 1270 | |||
| XM_017021245.1 | XP_016876734.1 | 7 | 724 | |||
| GenBank mRNA | AB209114.1 | - | 11 | 1258 | ||
| AF105020.1 | - | 11 | 1439 | |||
| AK023804.1 | - | 5 | 2027 | |||
| AK025962.1 | - | 1 | 55 | |||
| AY090480.1 | - | 11 | 1439 | |||
| BC031651.1 | - | 11 | 1326 | 5169145 | ||
| BM127609.1 | - | 5 | 360 | |||
| CR627447.1 | - | 11 | 1006 | |||

POMT2
Genomic Map