siRNA Details
| Gene Symbol: MFSD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152778.2 | NP_689991.1 | 2 | 185 | ||
| XM_005262893.1 | XP_005262950.1 | 1 | 99 | |||
| XM_005262896.1 | XP_005262953.1 | 1 | 99 | |||
| XM_005262897.2 | XP_005262954.1 | 1 | 99 | |||
| XM_005262898.2 | XP_005262955.1 | 1 | 99 | |||
| XM_011531830.1 | XP_011530132.1 | 1 | 99 | |||
| XM_011531831.1 | XP_011530133.1 | 1 | 99 | |||
| XM_011531832.1 | XP_011530134.1 | 1 | 99 | |||
| XM_017007989.1 | XP_016863478.1 | 1 | 99 | |||
| GenBank mRNA | AK074564.1 | - | 1 | 34 | ||
| AK315596.1 | - | 1 | 31 | |||
| BC029503.1 | - | 2 | 150 | 5272664 | ||

MFSD8
Genomic Map