siRNA Details
| Gene Symbol: CSF2RB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000395.2 | NP_000386.1 | 4 | 527 | ||
| XM_005261340.3 | XP_005261397.1 | 4 | 586 | |||
| XM_011529903.2 | XP_011528205.1 | 4 | 586 | |||
| XM_011529904.2 | XP_011528206.1 | 4 | 586 | |||
| XM_011529905.2 | XP_011528207.1 | 4 | 586 | |||
| GenBank mRNA | AK302993.1 | - | 2 | 171 | ||
| BC070085.1 | - | 4 | 527 | 30344148 | ||
| CR456428.1 | - | 3 | 383 | |||
| M59941.1 | - | 3 | 338 | |||

CSF2RB
Genomic Map