siRNA Details
Gene Symbol: KRTAP7-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181606.2 | NP_853637.2 | 1 | 538 | ||
GenBank mRNA | AJ457063.1 | - | 1 | 415 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KRTAP7-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181606.2 | NP_853637.2 | 1 | 538 | ||
GenBank mRNA | AJ457063.1 | - | 1 | 415 |