siRNA Details
Gene Symbol: KRTAP6-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181605.3 | NP_853636.3 | 1 | 423 | ||
GenBank mRNA | AB096951.1 | - | 1 | 423 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KRTAP6-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181605.3 | NP_853636.3 | 1 | 423 | ||
GenBank mRNA | AB096951.1 | - | 1 | 423 |