siRNA Details
| Gene Symbol: FAM151A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176782.2 | NP_788954.2 | 4 | 682 | ||
| XM_006710599.3 | XP_006710662.1 | 5 | 758 | |||
| GenBank mRNA | AK091901.1 | - | 4 | 711 | ||
| AY358629.1 | - | 4 | 668 | |||
| BC015993.1 | - | 4 | 684 | 4614435 | ||
| BC020874.1 | - | 4 | 680 | 4608844 | ||

FAM151A
Genomic Map