siRNA Details
| Gene Symbol: FAM151A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176782.2 | NP_788954.2 | 7 | 1092 | ||
| XM_006710599.3 | XP_006710662.1 | 8 | 1168 | |||
| GenBank mRNA | AK091901.1 | - | 7 | 1121 | ||
| AY358629.1 | - | 7 | 1078 | |||
| BC020874.1 | - | 7 | 1090 | 4608844 | ||

FAM151A
Genomic Map