siRNA Details
| Gene Symbol: NLRP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176820.3 | NP_789790.2 | 4 | 2171 | ||
| XM_011526894.2 | XP_011525196.1 | 2 | 298 | |||
| XM_011526895.1 | XP_011525197.1 | 2 | 203 | |||
| GenBank mRNA | AY154464.1 | - | 4 | 2143 | ||
| BC136583.1 | - | 4 | 2163 | 9020573 | ||

NLRP9
Genomic Map