siRNA Details
| Gene Symbol: CLEC4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080387.4 | NP_525126.2 | 6 | 782 | ||
| XM_011520632.2 | XP_011518934.1 | 7 | 751 | |||
| GenBank mRNA | AF411850.1 | - | 6 | 652 | ||
| AK314071.1 | - | 6 | 768 | |||
| AY115592.1 | - | 6 | 773 | |||
| AY297446.1 | - | 6 | 781 | |||
| AY486482.1 | - | 6 | 606 | |||
| AY486483.1 | - | 6 | 606 | |||
| BC032313.1 | - | 6 | 782 | 5218660 | ||

CLEC4D
Genomic Map