siRNA Details
| Gene Symbol: ANO9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012302.2 | NP_001012302.2 | 18 | 1694 | ||
| XM_011520053.2 | XP_011518355.1 | 18 | 1790 | |||
| XM_017017644.1 | XP_016873133.1 | 19 | 1824 | |||
| XM_017017645.1 | XP_016873134.1 | 19 | 1776 | |||
| XM_017017646.1 | XP_016873135.1 | 18 | 1713 | |||
| XM_017017647.1 | XP_016873136.1 | 18 | 1665 | |||
| GenBank mRNA | AK026923.1 | - | 18 | 1641 | ||
| AK074088.1 | - | 13 | 3476 | |||
| AK096874.1 | - | 9 | 982 | |||
| AL832289.1 | - | 8 | 883 | |||
| BC128557.1 | - | 18 | 1662 | 40122929 | ||
| BC144415.1 | - | 17 | 1720 | |||

ANO9
Genomic Map