siRNA Details
| Gene Symbol: HCAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177551.3 | NP_808219.1 | 1 | 493 | ||
| GenBank mRNA | AK290915.1 | - | 1 | 432 | ||
| AL698846.1 | - | 1 | 493 | |||
| AY148884.1 | - | 1 | 479 | |||
| BC027965.1 | - | 1 | 479 | 5222688 | ||
| BC056419.1 | - | 1 | 465 | 5747743 | ||

HCAR2
Genomic Map