siRNA Details
| Gene Symbol: HCAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177551.3 | NP_808219.1 | 1 | 576 | ||
| GenBank mRNA | AK290915.1 | - | 1 | 515 | ||
| AY148884.1 | - | 1 | 562 | |||
| BC027965.1 | - | 1 | 562 | 5222688 | ||
| BC056419.1 | - | 1 | 548 | 5747743 | ||

HCAR2
Genomic Map