siRNA Details
| Gene Symbol: KCNK18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181840.1 | NP_862823.1 | 2 | 282 | ||
| GenBank mRNA | AB087138.1 | - | 2 | 282 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNK18| Gene Symbol: KCNK18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181840.1 | NP_862823.1 | 2 | 282 | ||
| GenBank mRNA | AB087138.1 | - | 2 | 282 | ||