siRNA Details
| Gene Symbol: LUZP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009909.3 | NP_001009909.2 | 8 | 810 | ||
| NM_001252008.1 | NP_001238937.1 | 8 | 806 | |||
| NM_001252010.1 | NP_001238939.1 | 6 | 684 | |||
| XM_017017648.1 | XP_016873137.1 | 8 | 723 | |||
| GenBank mRNA | AK309077.1 | - | 6 | 459 | ||
| AL832641.3 | - | 8 | 722 | |||
| BC132829.1 | - | 8 | 602 | 40146851 | ||
| BC144177.1 | - | 8 | 598 | |||

LUZP2
Genomic Map