siRNA Details
| Gene Symbol: ANKRD13D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207354.2 | NP_997237.2 | 3 | 505 | ||
| NR_030767.1 | 4 | 588 | ||||
| XM_011544985.1 | XP_011543287.1 | 3 | 448 | |||
| XM_011544986.1 | XP_011543288.1 | 4 | 588 | |||
| XM_011544987.2 | XP_011543289.1 | 3 | 422 | |||
| GenBank mRNA | AK096277.1 | - | 3 | 418 | ||
| AK124721.1 | - | 4 | 585 | |||
| AK126438.1 | - | 2 | 502 | |||
| BC110419.1 | - | 3 | 478 | 5210878 | ||
| BC121024.1 | - | 3 | 379 | 40117420 | ||
| BC121025.2 | - | 3 | 379 | 40117423 | ||

ANKRD13D
Genomic Map