siRNA Details
| Gene Symbol: ANKRD13D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207354.2 | NP_997237.2 | 8 | 1039 | ||
| NR_030767.1 | 9 | 1122 | ||||
| XM_011544985.1 | XP_011543287.1 | 8 | 982 | |||
| XM_011544986.1 | XP_011543288.1 | 9 | 1122 | |||
| XM_011544987.2 | XP_011543289.1 | 8 | 956 | |||
| XM_011544988.2 | XP_011543290.1 | 3 | 302 | |||
| GenBank mRNA | AK027313.1 | - | 1 | 970 | ||
| AK057566.1 | - | 1 | 2496 | |||
| AK057681.1 | - | 1 | 1653 | |||
| AK096277.1 | - | 8 | 952 | |||
| AK124721.1 | - | 9 | 1119 | |||
| AK126438.1 | - | 7 | 1036 | |||
| BC110419.1 | - | 8 | 1012 | 5210878 | ||
| BC121024.1 | - | 8 | 913 | 40117420 | ||
| BC121025.2 | - | 8 | 913 | 40117423 | ||

ANKRD13D
Genomic Map