siRNA Details
| Gene Symbol: B4GALNT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178537.4 | NP_848632.2 | 10 | 984 | ||
| XM_017017654.1 | XP_016873143.1 | 10 | 2296 | |||
| GenBank mRNA | AB089939.1 | - | 10 | 975 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B4GALNT4| Gene Symbol: B4GALNT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178537.4 | NP_848632.2 | 10 | 984 | ||
| XM_017017654.1 | XP_016873143.1 | 10 | 2296 | |||
| GenBank mRNA | AB089939.1 | - | 10 | 975 | ||