siRNA Details
| Gene Symbol: KRT79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175834.2 | NP_787028.1 | 5 | 917 | ||
| GenBank mRNA | AJ564105.1 | - | 5 | 902 | ||
| AK130156.1 | - | 3 | 356 | |||
| BC039148.1 | - | 5 | 919 | 4746574 | ||
| BC063648.1 | - | 5 | 916 | 4804408 | ||

KRT79
Genomic Map