siRNA Details
| Gene Symbol: C12orf74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037671.3 | NP_001032760.1 | 2 | 867 | ||
| NM_001178097.2 | NP_001171568.1 | 2 | 867 | |||
| GenBank mRNA | BC043363.1 | - | 2 | 682 | ||
| BC043364.1 | - | 2 | 682 | |||
| BC105728.1 | - | 2 | 647 | 40025557 | ||
| BC107813.1 | - | 2 | 646 | 40025555 | ||

C12orf74
Genomic Map