siRNA Details
| Gene Symbol: RNASE10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012975.1 | NP_001012993.1 | 1 | 403 | ||
| GenBank mRNA | AK296766.1 | - | 4 | 891 | ||
| AY665805.1 | - | 1 | 403 | |||
| BC133002.1 | - | 1 | 422 | 40147024 | ||
| BC133028.1 | - | 1 | 422 | 40147050 | ||

RNASE10
Genomic Map