siRNA Details
Gene Symbol: TMEM202 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080462.1 | NP_001073931.1 | 2 | 250 | ||
XM_011521497.1 | XP_011519799.1 | 2 | 251 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMEM202 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080462.1 | NP_001073931.1 | 2 | 250 | ||
XM_011521497.1 | XP_011519799.1 | 2 | 251 |