siRNA Details
| Gene Symbol: ENPP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178543.4 | NP_848638.3 | 1, 2 | 296 | ||
| XM_011524737.1 | XP_011523039.1 | 1, 2 | 480 | |||
| GenBank mRNA | AK126250.1 | - | 1, 2 | 480 | ||
| AY230663.1 | - | 1, 2 | 279 | |||
| AY358622.1 | - | 1, 2 | 262 | |||
| BC041453.1 | - | 1, 2 | 279 | 5186743 | ||
| DB187443.1 | - | 1, 2 | 296 | |||

ENPP7
Genomic Map