siRNA Details
| Gene Symbol: CPLX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181654.3 | NP_857637.1 | 1 | 311 | ||
| GenBank mRNA | AB593158.1 | - | 1 | 292 | ||
| AK123184.1 | - | 1 | 311 | |||
| AY286502.1 | - | 1 | 141 | |||
| BC106887.1 | - | 1 | 304 | 40030305 | ||
| BC106888.1 | - | 1 | 304 | 40030309 | ||
| BX640655.1 | - | 1 | 290 | |||
| BX648518.1 | - | 1 | 294 | |||

CPLX4
Genomic Map