siRNA Details
| Gene Symbol: ZNF546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297763.1 | NP_001284692.1 | 7 | 1659 | ||
| NM_178544.4 | NP_848639.2 | 7 | 1737 | |||
| XM_011526898.2 | XP_011525200.1 | 6 | 2804 | |||
| XM_011526899.2 | XP_011525201.1 | 7 | 1649 | |||
| GenBank mRNA | AK122968.1 | - | 7 | 1631 | ||
| AK172809.1 | - | 1 | 206 | |||
| AK292528.1 | - | 7 | 1685 | |||
| BC045649.1 | - | 7 | 1651 | 5268795 | ||

ZNF546
Genomic Map