siRNA Details
Gene Symbol: TMEM240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114748.1 | NP_001108220.1 | 3 | 261 | ||
GenBank mRNA | BC147018.1 | - | 4 | 290 | 9056946 | |
BC147020.1 | - | 4 | 290 | 9056948 | ||
BU929336.1 | - | 3 | 242 |
