siRNA Details
| Gene Symbol: TFAP2E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178548.3 | NP_848643.2 | 7 | 1469 | ||
| XM_017001139.1 | XP_016856628.1 | 7 | 3768 | |||
| GenBank mRNA | AY326454.1 | - | 7 | 1469 | ||
| BC041175.1 | - | 7 | 1469 | 5786430 | ||

TFAP2E
Genomic Map