siRNA Details
| Gene Symbol: ZNF678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178549.3 | NP_848644.2 | 4 | 1852 | ||
| NR_033184.1 | 4 | 1880 | ||||
| GenBank mRNA | BC042500.1 | - | 4 | 1856 | 4823179 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF678| Gene Symbol: ZNF678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178549.3 | NP_848644.2 | 4 | 1852 | ||
| NR_033184.1 | 4 | 1880 | ||||
| GenBank mRNA | BC042500.1 | - | 4 | 1856 | 4823179 | |