siRNA Details
| Gene Symbol: CCDC190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178550.4 | NP_848645.3 | 4 | 732 | ||
| XM_005245125.3 | XP_005245182.1 | 4 | 3350 | |||
| XM_006711289.3 | XP_006711352.1 | 4 | 832 | |||
| XM_011509477.2 | XP_011507779.1 | 4 | 3353 | |||
| GenBank mRNA | AK123573.1 | - | 4 | 729 | ||

CCDC190
Genomic Map