siRNA Details
| Gene Symbol: LINC01139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015407.1 | 1 | 219 | |||
| GenBank mRNA | BC044843.1 | - | 1 | 219 | ||
| BC064529.1 | - | 1 | 225 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01139| Gene Symbol: LINC01139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015407.1 | 1 | 219 | |||
| GenBank mRNA | BC044843.1 | - | 1 | 219 | ||
| BC064529.1 | - | 1 | 225 | |||