siRNA Details
| Gene Symbol: ZFP69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320178.1 | NP_001307107.1 | 6 | 1477 | ||
| NM_001320179.1 | NP_001307108.1 | 6 | 1771 | |||
| NM_198494.2 | NP_940896.2 | 6 | 1474 | |||
| XM_005270809.4 | XP_005270866.1 | 6 | 2204 | |||
| XM_006710605.3 | XP_006710668.1 | 6 | 2184 | |||
| XM_006710606.3 | XP_006710669.1 | 6 | 2201 | |||
| GenBank mRNA | AK122618.1 | - | 6 | 1474 | ||
| BC041873.1 | - | 6 | 2187 | 5271835 | ||
| BC144174.1 | - | 5 | 1223 | |||

ZFP69
Genomic Map