siRNA Details
| Gene Symbol: SPOPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001664.2 | NP_001001664.1 | 2 | 463 | ||
| XM_005263655.4 | XP_005263712.1 | 2 | 329 | |||
| XM_005263656.4 | XP_005263713.1 | 2 | 329 | |||
| XM_005263657.4 | XP_005263714.1 | 2 | 339 | |||
| GenBank mRNA | BC071613.1 | - | 2 | 442 | 5275316 | |
| CN315400.1 | - | 2 | 466 | |||

SPOPL
Genomic Map