siRNA Details
| Gene Symbol: CYP27C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001665.3 | NP_001001665.3 | 3 | 509 | ||
| XM_017003960.1 | XP_016859449.1 | 4 | 699 | |||
| XM_017003961.1 | XP_016859450.1 | 2 | 400 | |||
| GenBank mRNA | AK131190.1 | - | 3 | 490 | ||
| BC039307.1 | - | 3 | 509 | 5266370 | ||

CYP27C1
Genomic Map