siRNA Details
| Gene Symbol: MROH2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287395.1 | NP_001274324.1 | 22 | 2523 | ||
| XM_011511075.2 | XP_011509377.1 | 22 | 2700 | |||
| XM_011511076.2 | XP_011509378.1 | 22 | 2700 | |||
| XM_011511077.2 | XP_011509379.1 | 22 | 2700 | |||
| XM_011511078.2 | XP_011509380.1 | 22 | 2661 | |||
| XM_011511079.2 | XP_011509381.1 | 22 | 2700 | |||
| XM_011511080.2 | XP_011509382.1 | 22 | 4171 | |||
| XM_011511081.2 | XP_011509383.1 | 22 | 2700 | |||
| XM_011511082.2 | XP_011509384.1 | 22 | 2700 | |||
| XM_011511083.2 | XP_011509385.1 | 22 | 2700 | |||
| XM_011511085.2 | XP_011509387.1 | 16 | 1770 | |||
| XM_011511086.2 | XP_011509388.1 | 22 | 2700 | |||

MROH2A
Genomic Map