siRNA Details
| Gene Symbol: PRR30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178553.3 | NP_848648.2 | 3 | 807 | ||
| GenBank mRNA | BC045671.1 | - | 3 | 753 | 5298697 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRR30| Gene Symbol: PRR30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178553.3 | NP_848648.2 | 3 | 807 | ||
| GenBank mRNA | BC045671.1 | - | 3 | 753 | 5298697 | |