siRNA Details
Gene Symbol: PRR30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178553.3 | NP_848648.2 | 3 | 838 | ||
GenBank mRNA | BC045671.1 | - | 3 | 784 | 5298697 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PRR30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178553.3 | NP_848648.2 | 3 | 838 | ||
GenBank mRNA | BC045671.1 | - | 3 | 784 | 5298697 |