siRNA Details
| Gene Symbol: MS4A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000139.4 | NP_000130.1 | 7 | 755 | ||
| NM_001256916.1 | NP_001243845.1 | 6 | 620 | |||
| XM_005273846.4 | XP_005273903.1 | 8 | 1070 | |||
| XM_011544850.2 | XP_011543152.1 | 8 | 688 | |||
| GenBank mRNA | BC074800.2 | - | 7 | 684 | 30915525 | |
| BC074843.2 | - | 7 | 684 | 30915222 | ||
| D10583.1 | - | 7 | 703 | |||
| JF411082.1 | - | 6 | 518 | |||

MS4A2
Genomic Map