siRNA Details
| Gene Symbol: KIAA2012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277372.2 | NP_001264301.2 | 22 | 3621 | ||
| XM_006712172.2 | XP_006712235.1 | 22 | 3626 | |||
| XM_011510467.2 | XP_011508769.1 | 22 | 3734 | |||
| XM_011510468.2 | XP_011508770.1 | 22 | 3629 | |||
| GenBank mRNA | AB095932.1 | - | 10 | 4129 | ||
| BC118982.1 | - | 10 | 1942 | 40026312 | ||

KIAA2012
Genomic Map